Merck
Cytotoxic thiocarbamate derivatives of boldine.
Natural product communications (2010-12-03)
Franz A Thomet, Pablo Pinyol, Joan Villena, Luis J Espinoza, Patricio G Reveco
ABSTRACT
Two new boldine derivatives: the 3-thiocarbamateboldine (3) and the 2,9-O,O-diacetyl-3-thiocarbamateboldine (4) have been synthesized and their cytotoxicity evaluated.
Product Description
Supelco
Boldine, analytical standard
Pricing and availability is not currently available.