Merck
  • Solvent effect on the optical property of uranyl acetylacetonate monohydrate.
Solvent effect on the optical property of uranyl acetylacetonate monohydrate.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2010-10-30)
Mukul Pradhan, Mrinmoyee Basu, Sougata Sarkar, Arun Kumar Sinha, Tarasankar Pal
ABSTRACT
The lability of the [UO2(acac)2H2O] complex has been exploited to decipher solvent composition of a medium. Successive blue shift of the π-π* band (λmax=282 nm) is observed due to alcohol substitution of increasing chain length in place of water. This observation helps to quantify the chain lengths of normal alcohol. The result has been accounted theoretically. However, in non-coordinating solvent, irregular red shift of the π-π* band is observed because of the molecular complexity. Again, charge transfer (CT) band at 211 nm has been identified employing polar-polar and polar coordinating-non-coordinating solvent systems.
Sigma-Aldrich
Manganese(III) acetylacetonate, technical grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Copper(II) acetylacetonate, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Copper(II) acetylacetonate, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chromium(III) acetylacetonate, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Manganese(II) acetylacetonate