Merck
  • Identification of human liver cytochrome P450 enzymes involved in the metabolism of SCH 530348 (Vorapaxar), a potent oral thrombin protease-activated receptor 1 antagonist.
Identification of human liver cytochrome P450 enzymes involved in the metabolism of SCH 530348 (Vorapaxar), a potent oral thrombin protease-activated receptor 1 antagonist.
Drug metabolism and disposition: the biological fate of chemicals (2010-10-12)
Anima Ghosal, Xiaowen Lu, Natalia Penner, Lan Gao, Ragu Ramanathan, Swapan K Chowdhury, Narendra S Kishnani, Kevin B Alton
ABSTRACT
Vorapaxar (SCH 530348), a potent oral thrombin protease-activated receptor 1 antagonist, is being developed as an antiplatelet agent for patients with established vascular disease. The objective of this study was to identify the human liver cytochrome P450 (P450) enzyme(s) responsible for the metabolism of SCH 530348. Human liver microsomes metabolized SCH 530348 to M19, an amine metabolite formed via carbamate cleavage, and M20 (monohydroxy-SCH 530348). Recombinant human CYP3A4 exhibited the most activity (11.5% profiled radioactivity) for the formation of M19, followed by markedly less substrate conversion with CYP1A1 and CYP2C19. Trace levels of M19, a major excreted human metabolite, were detected with CYP1A2, CYP3A5, and CYP4F3A. Formation of M19 by human liver microsomes was inhibited 89% by ketoconazole (IC(50), 0.73 μM), 34% by tranylcypromine, and 89% by anti-CYP3A4 monoclonal antibody. There was a significant correlation between the rate of M19 formation and midazolam 1'-hydroxylation (r = 0.75) or M19 formation and testosterone 6β-hydroxylation (r = 0.92). The results of screening, inhibition, and correlation studies confirmed that CYP3A4 is the major P450 enzyme responsible for M19 formation from SCH 530348. In contrast, formation of M20, a major circulating human metabolite at steady state, was primarily catalyzed by CYP3A4 and CYP2J2. M20 is pharmacologically equipotent to SCH 530348, whereas M19 is an inactive metabolite. Formation of M20 by human liver microsomes was inhibited 89% by ketoconazole, 75% by astemizole (a CYP2J2 inhibitor), and 43% by CYP3A4 monoclonal antibody. These results suggest that CYP3A4 and CYP2J2 are both involved in the formation of M20 metabolite.
Sigma-Aldrich
Astemizole, ≥98% (HPLC)