Merck
  • Synthesis, DNA binding, photo-induced DNA cleavage and cytotoxicity studies of europium(III) complexes.
Synthesis, DNA binding, photo-induced DNA cleavage and cytotoxicity studies of europium(III) complexes.
Dalton transactions (Cambridge, England : 2003) (2010-10-06)
Gong-Jun Chen, Xin Qiao, Jin-Lei Tian, Jing-Yuan Xu, Wen Gu, Xin Liu, Shi-Ping Yan
ABSTRACT
Two Eu(III) complexes, [Eu(acac)(3)(dpq)] (1) and [Eu(acac)(3)(dppz)] CH(3)OH (2) {viz. acetylacetonate (acac), dipyrido[3,2-d:20,30-f]quinoxaline (dpq), dipyrido[3,2-a:20,30-c] phenazine (dppz)}, have been synthesized and their DNA binding, photo-induced DNA cleavage activity and cell cytotoxicity are studied. The complexes display significant binding propensity to the calf thymus DNA in the order: 2(dppz) >1(dpq). Cleavage experiments using pBR322 supercoiled DNA suggest major groove binding for 2 and minor groove binding for 1. The mechanistic aspects on natural light (natural light in room during the day) and UV-A (365 nm) irradiation are via a mechanistic pathway involving formation of singlet oxygen and hydroxyl radical as the reactive species. The photo-induced DNA cleavage activity of 2 is also stronger than 1. The cytotoxicity of 1 and 2 against HeLa (cervical) cancer cells show that the IC(50) value of 19.11 ± 3.56 μM and 17.95 ± 5.47 μM, respectively.
Sigma-Aldrich
Manganese(II) acetylacetonate
Sigma-Aldrich
Chromium(III) acetylacetonate, 97%
Sigma-Aldrich
Copper(II) acetylacetonate, 97%
Sigma-Aldrich
Manganese(III) acetylacetonate, technical grade
Sigma-Aldrich
Copper(II) acetylacetonate, ≥99.9% trace metals basis