Merck
Towards preparative-scale, biocatalytic alkene reductions.
Chemical communications (Cambridge, England) (2010-10-06)
Despina J Bougioukou, Adam Z Walton, Jon D Stewart
ABSTRACT
Simple strategies for using alkene reductase enzymes to produce gram-scale quantities of both (R)- and (S)-citronellal have been developed. The methodology is easily accessible to non-specialist laboratories, allowing alkene reductases to be added to the toolbox of routine synthetic transformations.
Sigma-Aldrich
(S)-(−)-Citronellal, 96%
Sigma-Aldrich
(±)-Citronellal, natural, ≥85%, FCC, FG
Supelco
(±)-Citronellal, analytical standard
Sigma-Aldrich
(±)-Citronellal, ≥95.0% (GC)
Sigma-Aldrich
(R)-(+)-Citronellal, technical grade