MilliporeSigma
  • Dual inhibitory effect of Glycyrrhiza glabra (GutGard™) on COX and LOX products.
Dual inhibitory effect of Glycyrrhiza glabra (GutGard™) on COX and LOX products.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2010-09-25)
C V Chandrasekaran, H B Deepak, P Thiyagarajan, S Kathiresan, Gopal Krishna Sangli, M Deepak, Amit Agarwal
ABSTRACT
Glycyrrhiza glabra and its phytoconstituents have been known to possess widespread pharmacological properties as an anti-inflammatory, anti-viral, antitumour and hepatoprotective drug. In this study, we examined the inhibitory potential of extract of G. glabra (GutGard™) root and its phytoconstituents (glabridin, glycyrrhizin, and isoliquiritigenin) on both cyclooxygenase (COX) and lipoxygenase (LOX) products in order to understand the mechanism of its anti-inflammatory action. Inhibitory effect of GutGard™ and its phytoconstituents on lipopolysaccharide (LPS) induced prostaglandin E(2) (PGE(2)), calcimycin (A23187) induced thromboxane (TXB(2)), and leukotriene (LTB(4)) release was studied using murine macrophages (J774A.1) and human neutrophil (HL-60) cells. Results revealed that, G. glabra and glabridin significantly inhibited PGE(2), TXB(2) (COX) and LTB(4) (LOX), while, isoliquiritigenin exerted inhibitory effect only against COX products but failed to suppress LOX product. However, glycyrrhizin at the tested concentrations failed to exhibit inhibitory effect on both COX and LOX products. Here, we report for the first time that G. glabra (almost devoid of glycyrrhizin) exhibits anti-inflammatory property likely through the inhibition of PGE(2), TXB(2) and LTB(4) in mammalian cell assay system, which could be influenced in part by glabridin and isoliquiritigenin.
Sigma-Aldrich
Glabridin, ≥98% (HPLC), powder