Merck
  • Guar gum and scleroglucan interactions with borax: experimental and theoretical studies of an unexpected similarity.
Guar gum and scleroglucan interactions with borax: experimental and theoretical studies of an unexpected similarity.
The journal of physical chemistry. B (2010-09-25)
Gianfranco Bocchinfuso, Claudia Mazzuca, Chiara Sandolo, Silvia Margheritelli, Franco Alhaique, Tommasina Coviello, Antonio Palleschi
ABSTRACT
Guar gum is a galactomannan that assumes a very flexible conformation in solution, while Scleroglucan is a very rigid polysaccharide that dissolves in water as triple helices. Both polymers can form gels in the presence of borax. Despite their structural differences, the freeze-dried gel systems of both polymers, when compressed to form tablets, show a peculiar anisotropic swelling in water that reflects an amazing similarity in terms of their molecular properies. In this paper the behavior of the Guar/borax gel is compared with that of Scleroglucan/borax. The macroscopic properties of the two systems were characterized in terms of rheological measurements. Atomic force microscopy images and molecular dynamics simulation allowed to evaluate, at molecular level, the effect of borax addition to the Guar polymer. Both experiments show that an increasing of the polymer rigidity is produced by borax. The role played by galactose in the side chain was also discussed.
Sigma-Aldrich
Sodium tetraborate decahydrate, BioXtra, ≥99.5%
Sigma-Aldrich
Sodium tetraborate decahydrate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium tetraborate decahydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium tetraborate decahydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, 99.0-103.0%
Sigma-Aldrich
Sodium tetraborate decahydrate, puriss. p.a., ACS reagent, reag. ISO, buffer substance, ≥99.5%