Merck
  • Acidulant and oven type affect total anthocyanin content of blue corn cookies.
Acidulant and oven type affect total anthocyanin content of blue corn cookies.
Journal of the science of food and agriculture (2010-09-18)
Jian Li, Chuck E Walker, Jon M Faubion
ABSTRACT
Anthocyanins, pink to purple water-soluble flavonoids, are naturally occurring pigments with claimed health benefits. However, they are sensitive to degradation by high pH, light and temperature. Blue corn (maize) contains high levels of anthocyanins. Cookies are popular snacks and might serve as a vehicle to deliver antioxidants. A cookie formula with a high level of blue corn was developed with added acidulents and baked in ovens with different heat transfer coefficients. The best whole-grain blue corn flour/wheat pastry flour ratio (80:20 w/w), guar gum level (10 g kg(-1), flour weight basis) and water level (215 g kg(-1), flour weight basis) were determined based on response surface methodology analysis. The interactions of citric and lactic acids and glucono-δ-lactone with three oven types having different heat transfer coefficients (impingement oven 179 °C/4 min, reel oven 204 °C/10 min and convection oven 182 °C/4 min) influenced the total anthocyanin content (TAC) remaining in blue corn-containing cookies after baking. Cookies baked with citric acid in the convection oven retained the maximum TAC (227 ± 3 mg kg(-1)). By baking rapidly at lower temperatures and adding acidulents, it may be possible to increase residual natural source antioxidants in baked foods.
Sigma-Aldrich
D-(+)-Gluconic acid δ-lactone, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Gluconolactone, 99.0-101.0%, meets USP testing specifications
Pricing and availability is not currently available.