Merck
  • The humic acid analogue antraquinone-2,6-disulfonate (AQDS) serves as an electron shuttle in the electricity-driven microbial dechlorination of trichloroethene to cis-dichloroethene.
The humic acid analogue antraquinone-2,6-disulfonate (AQDS) serves as an electron shuttle in the electricity-driven microbial dechlorination of trichloroethene to cis-dichloroethene.
Bioresource technology (2010-08-17)
Federico Aulenta, Veronica Di Maio, Tommaso Ferri, Mauro Majone
ABSTRACT
Quinone moieties in humic substances have previously been shown to serve as extracellular electron acceptors in different metabolic pathways. Here we show that the humic acid analogue antraquinone-2,6-disulfonate (AQDS) can also serve as an electron donor in the microbial reductive dechlorination of TCE to cis-DCE. In a bioelectrochemical system (BES), equipped with a glassy carbon electrode (cathode) polarized at -250mV vs. SHE, electrically reduced AQDS served as the shuttle of electrons between the electrode surface and the dechlorinating bacteria. Interestingly, AQDS selectively stimulated only the first step of the TCE dechlorination sequence, leading to the formation of cis-DCE. Bioelectrochemical experiments carried out using a dechlorinating culture, highly enriched in the cis-DCE dechlorinating microorganism Dehalococcoides spp., confirmed the inability of reduced AQDS to serve as an electron donor for cis-DCE dechlorination. The results of this study have implications for the development of bioelectrochemical systems for groundwater remediation, as well as for the biogeochemical fate of chlorinated solvents in humic substances-rich subsurface environments.
Product Description
Supelco
cis-1,2-Dichloroethene solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
1,2-Dichloroethylene, mixture of cis and trans, 98%
Sigma-Aldrich
cis-1,2-Dichloroethylene, 97%
Sigma-Aldrich
trans-1,2-Dichloroethylene, 98%
Supelco
trans-1,2-Dichloroethylene, analytical standard
Supelco
cis-1,2-Dichloroethene, analytical standard