Merck
Fragrance material review on 2-ethyl-1-hexanol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2010-08-13)
D McGinty, J Scognamiglio, C S Letizia, A M Api
ABSTRACT
A summary of the safety data available for 2-ethyl-1-hexanol when used as a fragrance ingredient is presented. 2-Ethyl-1-hexanol is a member of the fragrance structural group branched chain saturated alcohols in which the common characteristic structural element is one hydroxyl group per molecule, and a C(4) to C(12) carbon chain with one or several methyl side chains. This review contains a detailed summary of all available toxicology and dermatology papers that are related to this individual fragrance ingredient and is not intended as a stand-alone document. A safety assessment of the entire branched chain saturated alcohol group will be published simultaneously with this document; please refer to Belsito et al. (2010) for an overall assessment of the safe use of this material and all other branched chain saturated alcohols in fragrances.
Sigma-Aldrich
2-Ethyl-1-hexanol, puriss., ≥99.0% (GC)
Sigma-Aldrich
Titanium(IV) 2-ethylhexyloxide, 95%
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99.6%
Supelco
2-Ethyl-1-hexanol, analytical standard
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99%, FG