Merck
  • [Oxidation of dermatan sulfate with a NaOCl-NaBr-2,2,6,6-tetramethylpiperidine-1-oxyl reagent in the water medium].
[Oxidation of dermatan sulfate with a NaOCl-NaBr-2,2,6,6-tetramethylpiperidine-1-oxyl reagent in the water medium].
Bioorganicheskaia khimiia (2010-07-21)
I Iu Ponedel'kina, E A Khaĭbrakhmanova, V N Odinokov, L M Khalilov, U M Dzhemilev
ABSTRACT
The oxidation of primary hydroxyl groups in dermatan sulfate with the NaOCl-NaBr-2,2,6,6-tetramethylpiperidine-1-oxyl (TEMPO) reagent in an aqueous alkaline environment was carried out for the first time. Modified dermatan sulfates containing hydrated aldehyde (15-50%) and carboxyl (25-100%) groups were obtained.
Sigma-Aldrich
Sodium bromide, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Sodium bromide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium bromide, BioXtra, ≥99.0%
Supelco
Density Standard 1251 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Sodium bromide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium bromide, ACS reagent, ≥99.0%