Merck
  • Development and evaluation of a 68Ga labeled pamoic acid derivative for in vivo visualization of necrosis using positron emission tomography.
Development and evaluation of a 68Ga labeled pamoic acid derivative for in vivo visualization of necrosis using positron emission tomography.
Bioorganic & medicinal chemistry (2010-06-29)
Kristof Prinsen, Junjie Li, Hubert Vanbilloen, Peter Vermaelen, Ellen Devos, Luc Mortelmans, Guy Bormans, Yicheng Ni, Alfons Verbruggen
ABSTRACT
In this study, we labeled N,N'-bis(diethylenetriamine pentaacetic acid)-pamoic acid bis-hydrazide (bis-DTPA-PA) with the generator produced PET radionuclide gallium-68 and evaluated 68Ga-bis-DTPA-PA as a potential tracer for in vivo visualization of necrosis by positron emission tomography (PET). Radiolabeling was achieved with a decay-corrected radiochemical yield of 63%. Biodistribution and in vivo stability studies in normal mice showed that 68Ga-bis-DTPA-PA is cleared faster from normal tissue than the previously reported 99mTc(CO)3 complex with bis-DTPA-PA which on the other hand is more stable in vivo. 68Ga-bis-DTPA-PA showed a 3.5-5 times higher binding to necrotic tissue than to viable tissue as shown by in vitro autoradiography while no statistically significant increased hepatic uptake was found in a biodistribution study in a mouse model of hepatic apoptosis. Specificity and avidity for necrosis was further evaluated in rats with a reperfused partial liver infarction and ethanol induced muscular necrosis. Dynamic microPET images showed a fast and prolonged uptake of 68Ga-bis-DTPA-PA in necrotic tissue with in vivo and ex vivo images correlating well with histochemical stainings. With necrotic to viable tissue activity ratios of 8-15 on ex vivo autoradiography, depending on the necrosis model, 68Ga-bis-DTPA-PA showed a faster and higher uptake in necrotic tissue than the 99mTc(CO)3 analog. These results show that 68Ga-bis-DTPA-PA specifically binds to necrotic tissue and is a promising tracer for in vivo visualization of necrosis using PET.
Sigma-Aldrich
Pamoic acid disodium salt