Merck
  • Simultaneous determination of residues of trichlorfon and dichlorvos in animal tissues by LC-MS/MS.
Simultaneous determination of residues of trichlorfon and dichlorvos in animal tissues by LC-MS/MS.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2010-05-01)
G M Wang, H Dai, Y G Li, X L Li, J Z Zhang, L Zhang, Y Y Fu, Z G Li
ABSTRACT
Trichlorfon is a thermally unstable insecticide that can be easily decomposed to dichlorvos at high temperatures. In this study, a liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was developed for simultaneous determination of the residues of trichlorfon and dichlorvos in animal tissues. Trichlorfon and dichlorvos in animal tissues were extracted with dichloromethane, homogenized/shaken, concentrated, and determined by LC-MS/MS. The two insecticides can be effectively separated and individually determined. The limits of detection (LODs) of trichlorfon and dichlorvos were 0.04 and 0.07 microg kg(-1), respectively; the limits of quantification (LOQs) of the two insecticides were both 5 microg kg(-1). The average recoveries for three spiked blank samples at 10, 20 and 40 microg kg(-1) were in the range 85-106%, with relative standard deviations (RSDs) below 10.6%, which falls in a reasonable range for the analysis of the two insecticides in animal-derived foods. The method is fit-for-purpose for the simultaneous determination of residues of trichlorfon and dichlorvos in animal tissues.
Product Description
Supelco
Trichlorfon, PESTANAL®, analytical standard
Pricing and availability is not currently available.