- Biomimetic synthesis and structural refinement of the macrocyclic dimer aminoglycoside 66-40C--the remarkably selective self-condensation of a putative aldehyde intermediate in the submerged culture medium producing sisomicin.
Biomimetic synthesis and structural refinement of the macrocyclic dimer aminoglycoside 66-40C--the remarkably selective self-condensation of a putative aldehyde intermediate in the submerged culture medium producing sisomicin.
Chemical communications (Cambridge, England) (2010-03-12)
Stephen Hanessian, Juan Pablo Maianti
PMID20221476
ABSTRACT
Aminoglycoside 66-40C, an unprecedented 16-membered bis-azadiene macrocyclic natural product isolated from the Micromonospora producer of the antibiotic sisomicin, was synthesized following a biomimetic strategy which definitively established its origin as arising from a remarkably selective non-enzymatic macro-dimerization.