- Determination of an inoculum effect with various cephalosporins among clinical isolates of methicillin-susceptible Staphylococcus aureus.
Determination of an inoculum effect with various cephalosporins among clinical isolates of methicillin-susceptible Staphylococcus aureus.
Antimicrobial agents and chemotherapy (2010-03-10)
Esteban C Nannini, Martin E Stryjewski, Kavindra V Singh, Tom H Rude, G Ralph Corey, Vance G Fowler, Barbara E Murray
PMID20211890
ABSTRACT
Using 98 clinical methicillin-susceptible Staphylococcus aureus isolates of known beta-lactamase (Bla) type, we found a pronounced inoculum effect for cephalexin (mostly Bla type A and C strains), a mild inoculum effect for cephalothin (especially types B and C), and no inoculum effects for ceftriaxone and cefuroxime. Ceftobiprole showed the lowest MICs at a high inoculum but with a slight increase for Bla-positive versus Bla-negative strains. Since a potential therapeutic effect associated with a cephalosporin inoculum effect has been described, further studies are warranted.
MATERIALS