Merck
N-acetylserotonin activates TrkB receptor in a circadian rhythm.
Proceedings of the National Academy of Sciences of the United States of America (2010-02-06)
Sung-Wuk Jang, Xia Liu, Sompol Pradoldej, Gianluca Tosini, Qiang Chang, P Michael Iuvone, Keqiang Ye
ABSTRACT
Brain-derived neurotrophic factor (BDNF) is a cognate ligand for the TrkB receptor. BDNF and serotonin often function in a cooperative manner to regulate neuronal plasticity, neurogenesis, and neuronal survival. Here we show that NAS (N-acetylserotonin) swiftly activates TrkB in a circadian manner and exhibits antidepressant effect in a TrkB-dependent manner. NAS, a precursor of melatonin, is acetylated from serotonin by AANAT (arylalkylamine N-acetyltransferase). NAS rapidly activates TrkB, but not TrkA or TrkC, in a neurotrophin- and MT3 receptor-independent manner. Administration of NAS activates TrkB in BDNF knockout mice. Furthermore, NAS, but not melatonin, displays a robust antidepressant-like behavioral effect in a TrkB-dependent way. Endogenous TrkB is activated in wild-type C3H/f(+/+) mice but not in AANAT-mutated C57BL/6J mice, in a circadian rhythm; TrkB activation is high at night in the dark and low during the day. Hence, our findings support that NAS is more than a melatonin precursor, and that it can potently activate TrkB receptor.
Sigma-Aldrich
N-Acetyl-5-hydroxytryptamine, ≥99% (HPLC), powder