Merck
  • Low-cost fluorimetric determination of radicals based on fluorogenic dimerization of the natural phenol sesamol.
Low-cost fluorimetric determination of radicals based on fluorogenic dimerization of the natural phenol sesamol.
Analytical chemistry (2010-01-29)
Yumi Makino, Seiichi Uchiyama, Ken-ichi Ohno, Hidetoshi Arakawa
ABSTRACT
A novel fluorimetric method for determining radicals using the natural phenol sesamol as a fluorogenic reagent is reported. In this assay, sesamol was reacted with aqueous radicals to yield one isomer of a sesamol dimer exclusively. The dimer emitted purple fluorescence near 400 nm around neutral pH, where it assumed the monoanionic form. This method was applied to the straightforward detection of radical nitric oxide (NO). The ready availability of sesamol should enable rapid implementation of applications utilizing this new assay, particularly in high-throughput analysis or screening.
Sigma-Aldrich
Anisole, anhydrous, 99.7%
Supelco
Anisole, analytical standard
Supelco
Anisole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Anisole, ≥99%, FCC, FG
Sigma-Aldrich
Anisole, ReagentPlus®, 99%