Merck
  • Interactions of diorganolead(IV) with 3-(2-thienyl)-2-sulfanylpropenoic acid and/or thiamine: chemical and in vitro and in vivo toxicological results.
Interactions of diorganolead(IV) with 3-(2-thienyl)-2-sulfanylpropenoic acid and/or thiamine: chemical and in vitro and in vivo toxicological results.
Inorganic chemistry (2010-01-22)
José S Casas, M Victoria Castaño, Agustín Sánchez, José Sordo, M Dolores Torres, María D Couce, Angeles Gato, Carmen Alvarez-Lorenzo, M Félix Camiña, Eduardo E Castellano
ABSTRACT
The reactions of PbR(2)(OAc)(2) (R = Me, Ph) with 3-(2-thienyl)-2-sulfanylpropenoic acid (H(2)tspa) in methanol or ethanol afforded complexes [PbR(2)(tspa)] that electrospray ionization-mass spectrometry (ESI-MS) and IR data suggest are polymeric. X-ray studies showed that [PbPh(2)(tspa)(dmso)] x dmso, crystallized from a solution of [PbPh(2)(tspa)] in dmso, is dimeric, and that [HQ](2)[PbPh(2)(tspa)(2)] (Q = diisopropylamine), obtained after removal of [PbPh(2)(tspa)] from a reaction including Q, contains the monomeric anion [PbPh(2)(tspa)(2)](2-). In the solid state the lead atoms are O,S-chelated by the tspa(2-) ligands in all these products, and in the latter two have distorted octahedral coordination environments. NMR data suggest that tspa(2-) remains coordinated to PbR(2)(2+) in solution in dmso. Neither thiamine nor thiamine diphosphate reacted with PbMe(2)(NO(3))(2) in D(2)O. Prior addition of H(2)tspa protected LLC-PK1 renal proximal tubule cells against PbMe(2)(NO(3))(2); thiamine had no statistically significant effect by itself, but greatly potentiated the action of H(2)tspa. Administration of either H(2)tspa or thiamine to male albino Sprague-Dawley rats dosed 30 min previously with PbMe(2)(NO(3))(2) was associated with reduced inhibition of delta-ALAD by the organolead compound, and with lower lead levels in kidney and brain, but joint administration of both H(2)tspa and thiamine only lowered lead concentration in the kidney.
Sigma-Aldrich
Diisopropylamine, ≥99.5%
Sigma-Aldrich
Diisopropylamine, purified by redistillation, 99.95%
Sigma-Aldrich
Diisopropylamine, puriss. p.a., ≥99.0% (GC)