Merck
  • Reaction products of gamma-tocopherol with (E)-4-oxo-2-nonenal in acidic acetonitrile.
Reaction products of gamma-tocopherol with (E)-4-oxo-2-nonenal in acidic acetonitrile.
Bioscience, biotechnology, and biochemistry (2010-01-09)
Fumie Saito, Satoshi Iwamoto, Ryo Yamauchi
ABSTRACT
gamma-Tocopherol was reacted with (E)-4-oxo-2-nonenal (ONE) at 37 degrees C in an acidic acetonitrile solution. The reaction products were isolated by reversed-phase high-performance liquid chromatography and their structures were characterized to be 5-substituted gamma-tocopherols: 5-(1-(furan-2-yl)pentyl)-gamma-tocopherol (1), 3-(1-butyl-4,5,7-trimethyl-7-(4,8,12-trimethyltridecyl)-8,9-dihydro-7H-furo[3,2-f]chromen-2yl)propanal (2), and 1-(1-butyl-4,5,7-trimethyl-7-(4,8,12-trimethyltridecyl)-8,9-dihydro-7H-furo[3,2-f]chromen-2-yl)-4-(furan-2-yl)octan-3-one (3). Compound 1 was the predominant product under the mild acidic conditions, whereas compounds 2 and 3 were relatively stable and accumulated during the reaction. Compound 1 was detected as the product when gamma-tocopherol and ONE were incubated in methyl linoleate in the presence of dicetyl phosphate. The results indicate that gamma-tocopherol may trap ONE under acidic conditions of lipid peroxidation.
Product Description
Supelco
trans-9,12-Octadecadienoic acid methyl ester, certified reference material, 10 mg/mL in heptane
Supelco
Methyl linoleate, analytical standard
Sigma-Aldrich
Methyl linolelaidate
Sigma-Aldrich
Methyl linoleate, ≥98% (GC)
Supelco
Methyl linolelaidate, analytical standard
Supelco
cis-9,cis-12-Octadecadienoic acid methyl ester, certified reference material, 10 mg/mL in heptane