Merck
  • Factors controlling alkylbenzene and tetrachloroethene desorption from municipal solid waste components.
Factors controlling alkylbenzene and tetrachloroethene desorption from municipal solid waste components.
Environmental science & technology (2009-12-31)
Jovita M Saquing, Lisa A Mitchell, Bingyan Wu, Travis B Wagner, Detlef R U Knappe, Morton A Barlaz
ABSTRACT
Desorption rates of toluene, o-xylene and tetrachloroethene from individual municipal solid waste components [high-density polyethylene (HDPE); poly(vinyl chloride) (PVC); office paper; newsprint; and rabbit food, a model food and yard waste] were determined. Effects of sorbent and sorbate properties, solvent composition (ultrapure water, acidogenic and methanogenic leachates), and contact time ("aging") on desorption rates were evaluated. Hydrophobic organic contaminant (HOC) desorption from PVC and HDPE could be described with a single-parameter polymer diffusion model. In contrast, a three-parameter, biphasic polymer diffusion model was required to describe HOC desorption rates from biopolymer composites. In general, HOC desorption rates from plastics were rapid for HDPE (D = 10(-10) cm(2)/s), a rubbery polymer, but slower for PVC (D = 10(-13)-10(-14) cm(2)/s), a glassy polymer. For biopolymer composites, a large fraction of sorbed HOCs was rapidly released (D(r) = 10(-9)-10(-10) cm(2)/s) while the remaining fraction desorbed slowly (D(s) = 10(-11)-10(-16) cm(2)/s). The toluene desorption rate from PVC was 1 order of magnitude faster in acidogenic leachate than in either ultrapure water or methanogenic leachate, a result that was primarily attributed to the plasticizing effect of volatile fatty acids in acidogenic leachate. For biopolymer composites, small increases in the slowly desorbing HOC fraction were observed with increasing aging time.
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Xylenes, reagent grade
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Xylenes, ACS reagent, ≥98.5% xylenes + ethylbenzene basis
Sigma-Aldrich
Xylenes, histological grade
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Supelco
o-Xylene, suitable for HPLC, 98%