Merck
  • Platinum nanoparticles: the crucial role of crystal face and colloid stabilizer in the diastereoselective hydrogenation of cinchonidine.
Platinum nanoparticles: the crucial role of crystal face and colloid stabilizer in the diastereoselective hydrogenation of cinchonidine.
Chemistry (Weinheim an der Bergstrasse, Germany) (2009-12-30)
Erik Schmidt, Wolfgang Kleist, Frank Krumeich, Tamas Mallat, Alfons Baiker
ABSTRACT
The preparation of stable metal nanoparticles requires a strong interaction between the (organic) stabilizer and the metal surface that might alter the catalytic properties. This behavior has been described as "poisoning" since the stabilizer normally decreases the catalytic activity due to site blocking. Here we show a striking influence of the stabilizer on the selectivity in the hydrogenation of cinchonidine (CD) over poly(acrylic acid) (PAA)-stabilized Pt nanoparticles with well-defined shape distributions. In the hydrogenation of the heteroaromatic ring of cinchonidine in toluene, the diastereomeric excess of the (S)-hexahydrocinchonidine increased upon increasing Pt{111}/Pt{100} ratio, but this distinct shape selectivity was observed only after the oxidative removal of PAA at 473 K. The use of the as-prepared nanoparticles inverted the major diastereomer to R, and this isomer was formed also in acetic acid. This striking change in the diastereoselectivity indicates that poly(acrylic acid), which remains on the Pt surface after preparation, interacts with CD during hydrogenation almost as strongly as the solvent acetic acid. The PAA stabilizer plays a dual role: it allows one to control the size and shape of the nanoparticles during their synthesis, and it affects the rate and diastereoselectivity of the hydrogenation of CD probably through a "surface-localized acidification".
Sigma-Aldrich
Cinchonidine, 96%