Merck
  • Characterisation of free and glycosidically bound odourant compounds of Aragonez clonal musts by GC-O.
Characterisation of free and glycosidically bound odourant compounds of Aragonez clonal musts by GC-O.
Analytica chimica acta (2009-12-17)
Goreti Botelho, Arlete Mendes-Faia, Maria Cristina Clímaco
ABSTRACT
To evaluate the potential aroma of Aragonez clonal red musts, several free and glycosidically bound odourant compounds were extracted. Then, the gas chromatography-olfactometry (GC-O) posterior intensity method was used to evaluate their odour intensity and the compounds were identified by gas chromatography-mass spectrometry (GC-MS). A group of eight sniffers evaluated free and bound fractions of Aragonez musts and perceived forty-three and twenty-two odourant peaks respectively. Furaneol (burnt sugar, candy-cotton) and vanillin (vanilla, sweet) were identified in both free and bound fractions of Aragonez musts, indicating their grape-derived origin. Principal component analysis (PCA) was applied to the posterior intensity method data and a relationship between the different odourant compound variables and the free fractions was established. Two principal components (PCs) were found which together explained 100% of the total variance. A large number of potentially important but yet unknown odourants was detected by the GC-O analysis.
Sigma-Aldrich
4-Hydroxy-2,5-dimethyl-3(2H)-furanone, ≥98%, FCC, FG
Sigma-Aldrich
4-Hydroxy-2,5-dimethyl-3(2H)-furanone, natural, ≥98%, FG
Sigma-Aldrich
4-Hydroxy-2,5-dimethyl-3(2H)-furanone, ≥99.0% (GC)
Supelco
4-Hydroxy-2,5-dimethyl-3(2H)-furanone, analytical standard