Merck
  • Microwave-assisted functionalization of carbon nanostructures in ionic liquids.
Microwave-assisted functionalization of carbon nanostructures in ionic liquids.
Chemistry (Weinheim an der Bergstrasse, Germany) (2009-10-23)
Ivan Guryanov, Francesca Maria Toma, Alejandro Montellano López, Mauro Carraro, Tatiana Da Ros, Guido Angelini, Eleonora D'Aurizio, Antonella Fontana, Michele Maggini, Maurizio Prato, Marcella Bonchio
ABSTRACT
The effect of microwave (MW) irradiation and ionic liquids (IL) on the cycloaddition of azomethine ylides to [60]fullerene has been investigated by screening the reaction protocol with regard to the IL medium composition, the applied MW power, and the simultaneous cooling of the system. [60]Fullerene conversion up to 98 % is achieved in 2-10 min, by using a 1:3 mixture of the IL 1-methyl-3-n-octyl imidazolium tetrafluoroborate ([omim]BF(4)) and o-dichlorobenzene, and an applied power as low as 12 W. The mono- versus poly-addition selectivity to [60]fullerene can be tuned as a function of fullerene concentration. The reaction scope includes aliphatic, aromatic, and fluorous-tagged (FT) derivatives. MW irradiation of IL-structured bucky gels is instrumental for the functionalization of single-walled carbon nanotubes (SWNTs), yielding group coverages of up to one functional group per 60 carbon atoms of the SWNT network. An improved performance is obtained in low viscosity bucky gels, in the order [bmim]BF(4)> [omim]BF(4)> [hvim]TF(2)N (bmim=1-methyl-3-n-butyl imidazolium; hvim=1-vinyl-3-n-hexadecyl imidazolium). With this protocol, the introduction of fluorous-tagged pyrrolidine moieties onto the SWNT surface (1/108 functional coverage) yields novel FT-CNS (carbon nanostructures) with high affinity for fluorinated phases.
Sigma-Aldrich
1,2-Dichlorobenzene, suitable for HPLC, 99%
Supelco
1,2-Dichlorobenzene, PESTANAL®, analytical standard
Sigma-Aldrich
1,2-Dichlorobenzene, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Dichlorobenzene, anhydrous, 99%