Merck
  • Preparation and synthetic application of partially protected brassinosteroids.
Preparation and synthetic application of partially protected brassinosteroids.
Steroids (2009-09-30)
Vladimir A Khripach, Vladimir N Zhabinskii, Yuliya Y Zhiburtovich, Galina V Ivanova, Olga V Konstantinova, Dmitrii V Tsavlovskii, Sybille Lorenz, Bernd Schneider
ABSTRACT
Preparation of partially protected brassinosteroids is achieved through the reaction of the source material (24-epicastasterone and 24-epibrassinolide) with diol-specific reagents (2,2-dimethoxypropane and methylboronic acid). The obtained products were shown to be useful synthetic intermediates for further preparation of minor representatives of this class of natural phytohormones (such as 3,24-diepicastasterone and 3-dehydro-24-epibrassinolide).
Sigma-Aldrich
2,2-Dimethoxypropane, reagent grade, 98%