Merck
  • Phenolic and enolic hydroxyl groups in curcumin: which plays the major role in scavenging radicals?
Phenolic and enolic hydroxyl groups in curcumin: which plays the major role in scavenging radicals?
Journal of agricultural and food chemistry (2009-09-10)
Jian-Ying Feng, Zai-Qun Liu
ABSTRACT
The aim of this work is to clarify the antioxidant abilities of phenolic and enolic hydroxyl groups in curcumin. 1,7-bis(4-benzyloxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione (BEC), 1,7-bis(4-hydroxy-3-methoxyphenyl)heptane-3,5-diol (OHC), 1,7-bis(4-hydroxy-3-methoxyphenyl)heptane-3,5-dione (THC), and 1,7-bis(3,4-dihydroxyphenyl)-1,6-heptadiene-3,5-dione (BDC) are synthesized to determine the antioxidant activities by using antiradical assays against 2,2'-diphenyl-1-picrylhydrazyl (DPPH) radical, galvinoxyl radical, and 2,2'-azinobis(3-ethylbenzothiazoline-6-sulfonate) cation radical (ABTS*+) and by protecting DNA and erythrocyte against 2,2'-azobis(2-amidinopropane hydrochloride) (AAPH) induced oxidation. The phenolic hydroxyl is the main group for curcumin to trap DPPH, galvinoxyl, and ABTS*+ radicals. The conjugative system between enolic and phenolic hydroxyl groups is beneficial for curcumin to protect erythrocytes against hemin-induced hemolysis and to protect DNA against AAPH-induced oxidation, but is not beneficial for curcumin to protect erythrocytes against AAPH-induced hemolysis. More hydroxyl groups enhance the antioxidant effectiveness of curcumin in the experimental systems employed herein. Therefore, curcumin acts as an antioxidant through the phenolic hydroxyl group.
Sigma-Aldrich
Galvinoxyl, free radical