Merck
  • Discovery of a novel avian neurosteroid, 7alpha-hydroxypregnenolone, and its role in the regulation of the diurnal rhythm of locomotor activity in Japanese quail.
Discovery of a novel avian neurosteroid, 7alpha-hydroxypregnenolone, and its role in the regulation of the diurnal rhythm of locomotor activity in Japanese quail.
General and comparative endocrinology (2009-04-14)
Kazuyoshi Tsutsui, Kazuhiko Inoue, Hitomi Miyabara, Saori Suzuki, Yuki Ogura, Yasuko Tobari, Shogo Haraguchi
ABSTRACT
The discovery of two novel avian neurosteroids in the quail brain, 7alpha- and 7beta-hydroxypregnenolone is described. Intracerebroventricular administration of 7alpha-hydroxypregnenolone, but not 7beta-hydroxypregnenolone was found to stimulate locomotor activity of male quail when spontaneous nocturnal activity is low. Diurnal changes in locomotor activity in male quail were found to be correlated with a diurnal change in the concentration of diencephalic 7alpha-hydroxypregnenolone. This correlation was a not seen in female quail which have a lower amplitude diurnal rhythm of locomotor activity and lower daytime concentrations of diencephalic 7alpha-hydroxypregnenolone. Treatment of male quail with melatonin was found to depress the synthesis of 7alpha-hydroxypregnenolone in the diencephalon. This is a previously undescribed role for melatonin in the regulation of neurosteroid synthesis in the brain of any vertebrate. We therefore deduced in male quail, that the nocturnal depression in locomotory activity is a consequence of a depression in diencephalic 7alpha-hydroxypregnenolone resulting from the inhibitory action of the nocturnal increase in melatonin. This observation may be of widespread significance for the molecular control of rhythmic locomotor activity in all vertebrates.