Merck
  • The biomimetic mineralization of double-stranded and cylindrical helical BaCO(3) nanofibres.
The biomimetic mineralization of double-stranded and cylindrical helical BaCO(3) nanofibres.
Chemical communications (Cambridge, England) (2009-02-20)
Jian-Hua Zhu, Shu-Hong Yu, An-Wu Xu, Helmut Cölfen
ABSTRACT
A facile biomimetic method is reported for the synthesis of novel BaCO(3) nanofibres with double-stranded and cylindrical helical morphologies via a phosphonated block co-polymer-controlled mineralization process.
Sigma-Aldrich
Barium carbonate, 99.98% trace metals basis
Sigma-Aldrich
Barium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Barium carbonate, ACS reagent, ≥99%