Merck
  • Occurrence, source diagnosis, and biological effect assessment of DDT and its metabolites in various environmental compartments of the Pearl River Delta, South China: a review.
Occurrence, source diagnosis, and biological effect assessment of DDT and its metabolites in various environmental compartments of the Pearl River Delta, South China: a review.
Environmental pollution (Barking, Essex : 1987) (2009-01-27)
Ying Guo, Huan-Yun Yu, Eddy Y Zeng
ABSTRACT
This review summarizes and analyzes available data in the literature (mostly after 2000) on the occurrence of dichlorodiphenyltrichloroethane (DDT) and its main metabolites, dichlorodiphenyldichloroethane (DDD) and chlorodiphenyldichloroethylene (DDE), in the environment of the Pearl River Delta (PRD), South China. Generally, the concentration levels of the sum of DDT, DDD, and DDE, designated as DDTs thereafter, have not significantly declined since 1983. However, the composition of DDTs residues has changed with time. DDTs in soil, freshwater sediment and freshwater fish species were mainly residues from chronological use. There is evidence that new point sources, such as dicofol and anti-fouling paint, may have contributed DDTs to various environmental compartments. Risk assessment against existing criteria indicated that the levels of DDTs in water and some fish species may pose adverse effects to humans or wildlife, and those in sediment/soil may also cause negative impacts to the eco-environment of the PRD.