Merck
  • Cyclisation of citronellal over heterogeneous inorganic fluorides--highly chemo- and diastereoselective catalysts for (+/-)-isopulegol.
Cyclisation of citronellal over heterogeneous inorganic fluorides--highly chemo- and diastereoselective catalysts for (+/-)-isopulegol.
Chemical communications (Cambridge, England) (2009-01-13)
Simona M Coman, Pratap Patil, Stefan Wuttke, Erhard Kemnitz
ABSTRACT
Based on a fluorolytic sol-gel synthesis, nanoscopic metal fluorides and partly hydroxylated metal fluorides were synthesized; varying the F : OH ratio inside these solids yielded catalysts with different combinations and variable strength Lewis and Brønsted acid sites, which demonstrated unexpected catalytic properties for the diastereoselective synthesis of (+/-)-isopulegol.
Sigma-Aldrich
(R)-(+)-Citronellal, technical grade
Supelco
(±)-Citronellal, analytical standard
Sigma-Aldrich
(±)-Citronellal, ≥95.0% (GC)
Sigma-Aldrich
(S)-(−)-Citronellal, 96%
Sigma-Aldrich
(±)-Citronellal, natural, ≥85%, FCC, FG