- Scar sarcoidosis with a 50-year interval between an accident and onset of lesions.
Scar sarcoidosis with a 50-year interval between an accident and onset of lesions.
Dermatology online journal (2008-12-20)
Hiram Larangeira de Almeida, Roberto Coswig Fiss
PMID19094856
ABSTRACT
Cutaneous sarcoidosis has been reported to develop in scars. We report an unusual case of sarcoidal granuloma formation in scars that had been present for 50 years.
MATERIALS