Merck
  • The topical niacin sensitivity test: an inter- and intra-rater reliability study in healthy controls.
The topical niacin sensitivity test: an inter- and intra-rater reliability study in healthy controls.
Prostaglandins, leukotrienes, and essential fatty acids (2008-07-29)
Melissa Kerr, Sue Cotton, Tina Proffitt, Mirabel McConchie, Connie Markulev, Stefan Smesny, Patrick McGorry, Gregor Berger
ABSTRACT
Topical application of nicotinic acid results in erythema, and in some cases oedema of the skin, supporting a strong relationship between niacin sensitivity and prostaglandin D2. The aim of this study was to examine the inter-rater and intra-rater reliability of a 12-min niacin sensitivity test in healthy adults. Three raters assessed the skin reaction of 12 volunteers, over 3-min intervals across four niacin concentrations (0.1, 0.01, 0.001, and 0.0001), and over six sessions. Inter-rater reliability estimates ranged from 0.85 to 0.97 for the total niacin sensitivity score. Similar inter-rater reliability estimates were found for niacin sensitivity ratings by concentration and time. Intra-rater reliability estimates ranged from 0.63 to 0.93 for the total niacin sensitivity score. These data indicate that the 12-min topical niacin sensitivity test has excellent reliability.
Sigma-Aldrich
Methyl nicotinate, ≥99%, FG
Sigma-Aldrich
Methyl nicotinate, 99%
Sigma-Aldrich
Methyl nicotinate, puriss., ≥99.0% (GC)