Merck
  • Responses to clomazone and 5-ketoclomazone by Echinochloa phyllopogon resistant to multiple herbicides in Californian rice fields.
Responses to clomazone and 5-ketoclomazone by Echinochloa phyllopogon resistant to multiple herbicides in Californian rice fields.
Pest management science (2008-05-22)
Hagai Yasuor, Patti L TenBrook, Ronald S Tjeerdema, Albert J Fischer
ABSTRACT
Late watergrass [Echinochloa phyllopogon (Stapf.) Koss.] is a major weed of Californian rice that has evolved P450-mediated metabolic resistance to multiple herbicides. Resistant (R) populations are also poorly controlled by the recently introduced herbicide clomazone. The authors assessed whether this cross-resistance was also P450 mediated, and whether R plants also had reduced sensitivity to photooxidation. Understanding mechanism(s) of resistance facilitates the design of herbicide management strategies to delay resistance evolution. Ratios (R/S) of R to susceptible (S) GR(50) were near 2.0. [(14)C]Clomazone uptake was similar in R and S plants. Clomazone and its metabolite 5-ketoclomazone reduced chlorophyll and carotenoids in S more than in R plants. The P450 inhibitors disulfoton and 1-aminobenzo-triazole (ABT) safened clomazone in R and S plants. Disulfoton safened 5-ketoclomazone only in S plants, while ABT synergized 5-ketoclomazone mostly against S plants. Paraquat was more toxic in S than in R plants. Cross-resistance to clomazone explains failures to control R plants in rice fields, and safening by P450 inhibitors suggests that oxidative activation of clomazone is needed for toxicity to E. phyllopogon. Clomazone resistance requires mitigation of 5-ketoclomazone toxicity, but P450 detoxification may not significantly confer resistance, as P450 inhibitors poorly synergized 5-ketoclopmazone in R plants. Responses to paraquat suggest research on mechanisms to mitigate photooxidation in R and S plants is needed.
Product Description
Supelco
Clomazone, PESTANAL®, analytical standard