Merck
  • Early oxidative damage underlying neurodegeneration in X-adrenoleukodystrophy.
Early oxidative damage underlying neurodegeneration in X-adrenoleukodystrophy.
Human molecular genetics (2008-03-18)
Stéphane Fourcade, Jone López-Erauskin, Jorge Galino, Carine Duval, Alba Naudi, Mariona Jove, Stephan Kemp, Francesc Villarroya, Isidre Ferrer, Reinald Pamplona, Manuel Portero-Otin, Aurora Pujol
ABSTRACT
X-linked adrenoleukodystrophy (X-ALD) is a fatal neurodegenerative disorder, characterized by progressive cerebral demyelination cerebral childhood adrenoleukodystrophy (CCALD) or spinal cord neurodegeneration (adrenomyeloneuropathy, AMN), adrenal insufficiency and accumulation of very long-chain fatty acids (VLCFA) in tissues. The disease is caused by mutations in the ABCD1 gene, which encodes a peroxisomal transporter that plays a role in the import of VLCFA or VLCFA-CoA into peroxisomes. The Abcd1 knockout mice develop a spinal cord disease that mimics AMN in adult patients, with late onset at 20 months of age. The mechanisms underlying cerebral demyelination or axonal degeneration in spinal cord are unknown. Here, we present evidence by gas chromatography/mass spectrometry that malonaldehyde-lysine, a consequence of lipoxidative damage to proteins, accumulates in the spinal cord of Abcd1 knockout mice as early as 3.5 months of age. At 12 months, Abcd1- mice accumulate additional proteins modified by oxidative damage arising from metal-catalyzed oxidation and glycoxidation/lipoxidation. While we show that VLCFA excess activates enzymatic antioxidant defenses at the protein expression levels, both in neural tissue, in ex vivo organotypic spinal cord slices from Abcd1- mice, and in human ALD fibroblasts, we also demonstrate that the loss of Abcd1 gene function hampers oxidative stress homeostasis. We find that the alpha-tocopherol analog Trolox is able to reverse oxidative lesions in vitro, thus providing therapeutic hope. These results pave the way for the identification of therapeutic targets that could reverse the deregulated response to oxidative stress in X-ALD.
Sigma-Aldrich
Hexacosanoic acid, technical, ≥90% (GC)
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Hexacosanoic acid, ≥95% (capillary GC)