Merck
  • Efficient synthesis of (+)-1,8,8a-tri-epi-swainsonine, (+)-1,2-di-epi-lentiginosine, (+)-9a-epi-homocastanospermine and (-)-9-deoxy-9a-epi-homocastanospermine from a D-glucose-derived aziridine carboxylate, and study of their glycosidase inhibitory activities.
Efficient synthesis of (+)-1,8,8a-tri-epi-swainsonine, (+)-1,2-di-epi-lentiginosine, (+)-9a-epi-homocastanospermine and (-)-9-deoxy-9a-epi-homocastanospermine from a D-glucose-derived aziridine carboxylate, and study of their glycosidase inhibitory activities.
Organic & biomolecular chemistry (2008-02-12)
K S Ajish Kumar, Vinod D Chaudhari, Dilip D Dhavale
ABSTRACT
The utility of a D-glucose-derived aziridine carboxylate was demonstrated for the synthesis of polyhydroxylated quinolizidine and indolizidine alkaloids. The chemoselective reduction of 1 followed by two-carbon homologation by the Wittig reaction afforded gamma,delta-aziridino-alpha,beta-unsaturated ester 9, which on regioselective nucleophilic aziridine ring opening either by using water as a nucleophile or hydrogenation afforded delta-lactams 11/16--true synthons for the synthesis of four structurally different iminosugars, namely quinolizidine alkaloids 5b/5c, swainsonine 6b and lentiginosine 7b analogues. Glycosidase inhibitory activities of these iminosugars were investigated.
Sigma-Aldrich
(1S,6S,7R,8R,8aR)-1,6,7,8-Tetrahydroxyindolizidine, 98%
Pricing and availability is not currently available.