Merck
  • Oxa-ene reaction of enols of amides with 4-phenyl-1,2,4-triazoline-3,5-dione.
Oxa-ene reaction of enols of amides with 4-phenyl-1,2,4-triazoline-3,5-dione.
The Journal of organic chemistry (2007-12-13)
Ahmad Basheer, Zvi Rappoport
ABSTRACT
The reaction of 16 enols of amides with 4-phenyl-1,2,4-triazoline-1,3-dione gave open chain adducts rather than the [2 + 2] cycloadducts with a hemiaminal moiety, both in solid state and in solution. This assignment is based on X-ray crystallography, (1)H and (13)C NMR data, and IR spectra. The suggested mechanism involves hydroxyl proton loss in a formal oxa-ene reaction. Mechanistic details and a possible alternative are discussed.
Sigma-Aldrich
4-Phenyl-1,2,4-triazoline-3,5-dione, 97%