Merck
Fragrance material review on ethyl cinnamate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-27)
S P Bhatia, G A Wellington, J Cocchiara, J Lalko, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of ethyl cinnamate when used as a fragrance ingredient is presented.
Sigma-Aldrich
Ethyl cinnamate, 99%
Sigma-Aldrich
Ethyl cinnamate, natural, ≥95%, FG
Sigma-Aldrich
Ethyl cinnamate, ≥98%, stabilized, FCC, FG