Merck
  • Versatile direct dehydrative approach for diaryliodonium(III) salts in fluoroalcohol media.
Versatile direct dehydrative approach for diaryliodonium(III) salts in fluoroalcohol media.
Chemical communications (Cambridge, England) (2007-10-11)
Toshifumi Dohi, Motoki Ito, Koji Morimoto, Yutaka Minamitsuji, Naoko Takenaga, Yasuyuki Kita
ABSTRACT
We have found that the use of fluoroalcohol media greatly enhanced the efficiency and scope of the direct dehydrative condensation of arenes and hypervalent iodine(III) compounds; the present clean method has a broad range of applicability as well as unique selectivity in the aromatic substrates, and is highly efficient even in polymer functionalization.
Product Description
Supelco
Mesitylene, analytical standard
Sigma-Aldrich
Mesitylene, reagent grade, 97%
Sigma-Aldrich
Mesitylene, 98%