- Medium-sized carbocycles by a zirconocene-catalyzed tandem formal ring expansion-magnesation reaction of alkenyl-substituted cyclic enol ethers.
Medium-sized carbocycles by a zirconocene-catalyzed tandem formal ring expansion-magnesation reaction of alkenyl-substituted cyclic enol ethers.
Organic letters (2007-07-20)
José Barluenga, Lucía Alvarez-Rodrigo, Félix Rodríguez, Francisco J Fañanás
PMID17637021
ABSTRACT
A new regio- and stereoselective zirconocene-catalyzed reaction for the synthesis of medium-sized rings is described. The global reaction supposes a formal ring expansion of a cyclic enol ether to give a functionalized carbocycle.