Merck
  • 1,1,2,2-tetrachloroethane reactions with OH-, Cr(II), granular iron, and a copper-iron bimetal: insights from product formation and associated carbon isotope fractionation.
1,1,2,2-tetrachloroethane reactions with OH-, Cr(II), granular iron, and a copper-iron bimetal: insights from product formation and associated carbon isotope fractionation.
Environmental science & technology (2007-07-07)
Martin Elsner, David M Cwiertny, A Lynn Roberts, Barbara Sherwood Lollar
ABSTRACT
Despite widespread implementation of zero-valent iron remediation schemes, the manner and order of chemical bond cleavage in iron-mediated organohalide transformations remains imperfectly understood. We present insights from carbon isotope fractionation for the dehalogenation of 1,1,2,2-tetrachloroethane (1,1,2,2-TeCA) and 1,1,1-trichloroethane (1,1,1-TCA) by various reactants. Elimination of HCl by OH- gave isotope fractionation in 1,1,2,2-TeCA of Euro = -25.6 per thousand, KIE(c) = 1.02 to 1.03 per carbon center, consistentwith a concerted (E2) mechanism. In contrast, 1,1,1-TCA reduction by Cr(II), Fe(0), and Cu-plated iron (Cu/Fe) resulted in Euro = -13.6 per thousand to -15.8 per thousand indicating the initial involvement of a single C-Cl bond (KIE(c) approximately 1.03). 1,1,2,2-TeCA reduction by Cr(II), Fe(0), and Cu/Fe yielded Euro = -18.7 per thousand, -19.3 per thousand, and -17.0 per thousand, respectively. In the two latter cases, depletion of the minor product TCE by 26 per thousand indicated its formation via nonreductive dehydrohalogenation. The major 1,1,2,2-TeCA reduction products, cis- and trans-DCE, differed by 2.3 per thousand +/- 1.0 per thousand in Cr(II) systems, but were equivalent in Fe(0) and Cu/Fe systems. In contrast, the ratio of cis-DCE to trans-DCE concentration was 2.5 for reduction with Cr(II) and Fe(0), but -3.8 with Cu/Fe. Complementary isotope and concentration data therefore suggest differences in the transition state geometry and/ or reaction intermediates in each reductant system.
Sigma-Aldrich
1,1,2,2-Tetrachloroethane, reagent grade, ≥98%