Merck
  • Et2Zn-catalyzed intramolecular hydroamination of alkynyl sulfonamides and the related tandem cyclization/addition reaction.
Et2Zn-catalyzed intramolecular hydroamination of alkynyl sulfonamides and the related tandem cyclization/addition reaction.
The Journal of organic chemistry (2007-06-22)
Yan Yin, Wenying Ma, Zhuo Chai, Gang Zhao
ABSTRACT
Intramolecular hydroamination of alkynyl amides was effected by a catalytic amount of Et2Zn (20 mol %) to form indole derivatives, and a tandem cyclization/nucleophilic addition procedure involving reaction of the indole zinc salt intermediate with acid chlorides or halides was developed to provide an efficient approach to C3-substituted indole derivatives when an excess of Et2Zn (120 mol %) was used.
Sigma-Aldrich
Diethylzinc solution, 1.0 M in hexanes
Sigma-Aldrich
Diethylzinc solution, 1.0 M in heptane
Sigma-Aldrich
Diethylzinc solution, 15 wt. % in toluene