Merck
  • Asymmetric amplification in catalysis by trans-1,2-diaminocyclohexane bistriflamide.
Asymmetric amplification in catalysis by trans-1,2-diaminocyclohexane bistriflamide.
Organic letters (2007-01-16)
Tummanapalli Satyanarayana, Benoit Ferber, Henri B Kagan
ABSTRACT
A strong asymmetric amplification is observed in the addition of diethylzinc on aromatic aldehydes in the presence of the bistriflamide of trans-1,2-diaminocyclohexane 3a. The asymmetric amplification originates from the insolubility of the catalyst precursor 3a of low enantiomeric excess (ee), with a concomitant large increase of ee for the minor soluble part of 3a. Controlled mono-N-acetylation of 3a (20% ee) at -78 degrees C allowed isolation of 4 possessing 90% ee. [reaction: see text].
Sigma-Aldrich
Diethylzinc solution, 15 wt. % in toluene
Sigma-Aldrich
Diethylzinc solution, 1.0 M in hexanes
Sigma-Aldrich
Diethylzinc solution, 1.0 M in heptane