Merck
  • Formation of CdS nanoparticle necklaces with functionalized dendronized polymers.
Formation of CdS nanoparticle necklaces with functionalized dendronized polymers.
Small (Weinheim an der Bergstrasse, Germany) (2006-12-29)
Yanhong Zhang, Yongming Chen, Haijun Niu, Mingyuan Gao
ABSTRACT
A one-dimensional assembly of cadmium sulfide (CdS) nanoparticles was prepared by using functionalized polymers with pendant Fréchet-type dendritic wedges of different generation. The novel dendronized polymers acted as both nanoreactors for the formation of CdS nanoparticles and a template to arrange the nanoparticles into a necklace. The polymers of different dendron generation were applied to stabilize CdS particles and the effect of dendron size was observed. CdS nanoparticle necklaces were obtained successfully when polymers bearing second-generation dendrons were applied.
Product Description
Cadmium(II) acetate, SAFC Hitech®, anhydrous, 99.995%
Sigma-Aldrich
Cadmium acetate dihydrate, reagent grade, 98%
Sigma-Aldrich
Cadmium acetate dihydrate, purum p.a., ≥98.0% (KT)
Sigma-Aldrich
Cadmium(II) acetate, anhydrous, 99.995%