Merck
  • Daphnia magna and ecotoxicogenomics: gene expression profiles of the anti-ecdysteroidal fungicide fenarimol using energy-, molting- and life stage-related cDNA libraries.
Daphnia magna and ecotoxicogenomics: gene expression profiles of the anti-ecdysteroidal fungicide fenarimol using energy-, molting- and life stage-related cDNA libraries.
Chemosphere (2006-11-23)
Anneleen Soetaert, Karlijn van der Ven, Lotte N Moens, Tine Vandenbrouck, Piet van Remortel, Wim M De Coen
ABSTRACT
In the present study, the existing life stage-specific cDNA library was extended with energy- and molting-related genes using Suppression Subtractive Hybridization PCR and a microarray for the aquatic test organism Daphnia magna was created. A gene set of 2455 fragments was produced belonging to different pathways such as carbohydrate and lipid metabolism, O2 transport and heme metabolism, immune response, embryo development, cuticula metabolism and visual perception pathways. Using this custom microarray, gene expression profiles were generated from neonates exposed to three concentrations of the anti-ecdysteroidal fungicide fenarimol (0.5, 0.75, 1 microg/ml) during 48 h and 96 h. In total, 59 non-redundant genes were differentially expressed, of which more genes were down- than up-regulated. The gene expression data indicated a main effect on molting specific pathways. At the highest concentration, a set of proteolytic enzymes - including different serine proteases and carboxypeptidases - were induced whereas different cuticula proteins were down-regulated (48 h). Moreover, effects on embryo development were demonstrated at the gene expression as well as at the organismal level. The embryo development related gene vitellogenin was differentially expressed after 96 h of exposure together with a significant increase in embryo abnormalities in the offspring. This study suggests that this Daphnia magna microarray is of great further value for the elucidation of molecular mechanisms of toxicity and for the future development of specific biomarkers for hazard characterization.