- Conversion of indanone oximes into isocarbostyrils.
Conversion of indanone oximes into isocarbostyrils.
Bioorganic & medicinal chemistry letters (2006-10-27)
Yasuhiro Torisawa, Shinji Aki, Jun-ichi Minamikawa
PMID17064893
ABSTRACT
New and effective method for the Beckmann rearrangement of indanone oxime mesylate is described, in which a selective and controlled production of the isomeric isocarbostyrils is achieved.
MATERIALS