Merck
  • Pyrene-appended alpha-cyclodextrin as a fluorescent pH probe responding to a wide range.
Pyrene-appended alpha-cyclodextrin as a fluorescent pH probe responding to a wide range.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2006-06-14)
Iwao Suzuki, Mihoko Ui, Akiyo Yamauchi
ABSTRACT
Pyrene-appended alpha-cyclodextrin (3) in which a trimethylenediamine linker connected the pyrene residue to the alpha-cyclodextrin moiety showed pH-dependent fluorescence intensity changes. The fluorescence intensity was almost linearly changed within the pH range of 5 - 10. The unique fluorescence response of 3 to the pH was due not only to the favorable pK(a) values (pK(a1) = 6.4 and pK(a2) = 8.8), but also to the almost equal contributions of the amino groups to the pyrene's fluorescence quenching.
Sigma-Aldrich
1,3-Diaminopropane dihydrochloride, 98%
Sigma-Aldrich
1,3-Diaminopropane, ≥99%