Merck
  • Survey of breakfast and infant cereals for aflatoxins B1, B2, G1 and G2.
Survey of breakfast and infant cereals for aflatoxins B1, B2, G1 and G2.
Food additives and contaminants (2006-06-06)
J Tam, M Mankotia, M Mably, P Pantazopoulos, R J Neil, P Calway, P M Scott
ABSTRACT
Three hundred and forty-nine breakfast and infant cereal samples were collected at retail level across Canada from 2002 to 2005. They included rice-, soy-, barley-based and mixed-grain infant cereals, corn-, wheat-, rice-based and mixed-grain breakfast cereals, and were analysed for aflatoxins B1, B2, G1 and G2 using a modified AOAC International official method. An immunoaffinity column was used for the cleanup and purification of extracts. Determination of aflatoxins was by LC using post-column derivatization with pyridinium hydrobromide perbromide and fluorescence detection. Results indicated that 50% of both breakfast and infant cereals had detectable levels (limit of detection = 0.002 ng g-1) of aflatoxin B1, which is the most toxic of the four toxins. The levels found varied from 0.002 to 1.00 ng g-1 for aflatoxin B1, from 0.002 to 0.14 ng g-1 for aflatoxin B2, from 0.008 to 0.27 ng g-1 for aflatoxin G1, and from 0.008 to 0.048 ng g-1 for aflatoxin G2. Only 4% of the breakfast cereals and 1% of the infant cereals had aflatoxin B1 levels exceeding 0.1 ng g-1, which is the European Union maximum limit for aflatoxin B1 in baby foods and processed cereal-based foods for infants and young children.
Sigma-Aldrich
Aflatoxin G2
Aflatoxin G2 solution, 3.80 μg/g in acetonitrile, ERM®, certified reference material