Merck
  • Mark-recapture without estimating population sizes: a tool to evaluate termite baits.
Mark-recapture without estimating population sizes: a tool to evaluate termite baits.
Bulletin of entomological research (2006-03-25)
M W J Crosland, N-Y Su
ABSTRACT
The mark-recapture technique is currently being used by many termite researchers. Here its two uses are explained and disentangled. Use 1 is mapping the geographical location of the colony and use 2 is estimating the number of termites in the colony. Use 2 is controversial because the accuracy of the estimates has been widely questioned. In this article, it is explained that use 1 is not controversial and that use 1 (not use 2) is the criterion that is currently being used for evaluating the efficacy of termite baits.
Sigma-Aldrich
Sudan Red 7B, for microscopy (Bot., Hist.)
Sigma-Aldrich
Sudan Red 7B
Supelco
Sudan Red 7B, analytical standard
Supelco
Hexaflumuron, PESTANAL®, analytical standard