Merck
  • Oxygen plasma surface modification enhances immobilization of simvastatin acid.
Oxygen plasma surface modification enhances immobilization of simvastatin acid.
Biomedical research (Tokyo, Japan) (2006-03-18)
Masao Yoshinari, Tohru Hayakawa, Kenichi Matsuzaka, Takashi Inoue, Yutaka Oda, Masaki Shimono, Takaharu Ide, Teruo Tanaka
ABSTRACT
Simvastatin acid (SVA) has been reported to stimulate bone formation with increased expression of BMP-2. Therefore, immobilization of SVA onto dental implants is expected to promote osteogenesis at the bone tissue/implant interface. The aim of this study was to evaluate the immobilization behavior of SVA onto titanium (Ti), O(2)-plasma treated titanium (Ti + O(2)), thin-film coatings of hexamethyldisiloxane (HMDSO), and O(2)-plasma treated HMDSO (HMDSO + O(2)) by using the quartz crystal microbalance-dissipation (QCM-D) technique. HMDSO surfaces were activated by the introduction of an OH group and/or O(2)-functional groups by O(2)-plasma treatment. In contrast, titanium surfaces showed no appreciable compositional changes by O(2)-plasma treatment. The QCM-D technique enabled evaluation even at the adsorption behavior of a substance with a low molecular weight such as simvastatin. The largest amount of SVA was adsorbed on O(2)-plasma treated HMDSO surfaces compared to untreated titanium, HMDSO-coated titanium, and O(2)-plasma treated titanium. These findings suggested that the adsorption of SVA was enhanced on more hydrophilic surfaces concomitant with the presence of an OH group and/or O(2)-functional group resulting from the O(2)-plasma treatment, and that an organic film of HMDSO followed by O(2)-plasma treatment is a promising method for the adsorption of SVA in dental implant systems.
Sigma-Aldrich
Hexamethyldisiloxane, ≥98%
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
Hexamethyldisiloxane, NMR grade, ≥99.5%
Sigma-Aldrich
Hexamethyldisiloxane, puriss., ≥98.5% (GC)