Merck
  • Development of a modified positive selection medium that allows to isolate Aspergillus oryzae strains cured of the integrated niaD-based plasmid.
Development of a modified positive selection medium that allows to isolate Aspergillus oryzae strains cured of the integrated niaD-based plasmid.
Bioscience, biotechnology, and biochemistry (2005-12-27)
Kazutomo Ishi, Taisuke Watanabe, Praveen Rao Juvvadi, Jun-Ichi Maruyama, Katsuhiko Kitamoto
ABSTRACT
The nitrate reductase gene (niaD) is the most frequently utilized as a selectable marker for homologous integration at the niaD locus of Aspergillus oryzae. In this study we developed a method for curing of the niaD-based plasmid integrated on the A. oryzae genome. Positive selection using a modified chlorate medium containing leucine as a nitrogen source enabled efficient isolation of the strains deficient in nitrate assimilation from the niaD(+) transformant. PCR analysis of the strains confirmed that the homologously integrated plasmid carrying the h2b-egfp fusion gene was cured by intrachromosomal recombination which was accompanied by the loss of the EGFP-fluorescence.
Product Description
Millipore
Potassium chlorate Supplement, suitable for microbiology
Sigma-Aldrich
Potassium chlorate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Potassium chlorate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium chlorate, puriss., 99-101%
Sigma-Aldrich
Potassium chlorate, ACS reagent, ≥99.0%