Merck
An unusual adverse drug reaction?
Allergologia et immunopathologia (2005-07-28)
J Montoro, N Freixenet, A Lozano, F Bertomeu
ABSTRACT
Adult Still Disease (ASD) is a rare (incidence 1-1,6/1,000,000 in Europe) seronegative polyarthropathy diagnosed with clinical criteria, excluding other etiologies. Minocycline, a semisynthetic derivative of tetracycline, has been associated with many adverse effects. We present the case of a 18-years-old man with a high suspicion of an adverse drug reaction (ADR), that was finally diagnosed of ASD. In order to exclude other diseases, laboratory test, radiology and allergological studies were performed. The results of the allergological studies excluded the possibility of an ADR. The rest of the results determined the diagnosis of ASD. It's necessary to emphasize the importance of an accurate differential diagnosis in cases like this, because many diseases may mimic an ADR, and may be underdiagnosed (or misdiagnosed).
Product Description
Dexchlorpheniramine maleate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
S-(+)-Chlorpheniramine maleate salt
Supelco
Chlorpheniramine Maleate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(±)-Chlorpheniramine maleate salt, ≥99% (perchloric acid titration)
Chlorphenamine maleate, European Pharmacopoeia (EP) Reference Standard